Valerenic acid ≥99% (by HPLC), powder
Supplier: INDOFINE CHEMICAL COMPANY MS
023801S10MG
100263-620EA
366.15
USD
100263-620
Valerenic acid ≥99% (by HPLC), powder
Valerenic acid
- White powder
- Store at 2-8 °C
Formula:
C₁₅H₂₂O₂ Melting Pt: 138…140 °C |
MDL Number:
MFCD00075694 CAS Number: 3569-10-6 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...