Triarachidin ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
Glycerol Trieicosanoate
, Triarachin
, 1,2,3-Trieicosanoyl Glycerol
332000100MG
100290-454EA
302.08
USD
100290-454
Triarachidin ≥99%
Triarachidin
Formula:
C₆₃H₁₂₂O₆ MW: 975.66 g/mol |
MDL Number:
MFCD00042879 CAS Number: 620-64-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...