Methyl perfluorohexadecanoate ≥95%
Supplier: INDOFINE CHEMICAL COMPANY MS
098895GM
100283-844EA
122.92
USD
100283-844
Methyl perfluorohexadecanoate ≥95%
Methyl perfluorohexadecanoate
Formula:
C₁₇H₃F₃₁O₂ |
ADR:
, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...