7-Methoxyisoflavone ≥99%, powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
7-Methoxy-3-phenyl-4H-chromen-4-one
1959610MG
100264-220EA
75.85
USD
100264-220
7-Methoxyisoflavone ≥99%, powder
7-Methoxyisoflavone
- Off white shining powder
- Store in a cool, dry place
Formula:
C₁₆H₁₂O₃ MW: 252.27 g/mol Melting Pt: 156.6…158.8 °C |
MDL Number:
MFCD00181416 CAS Number: 1621-56-3 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...