(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochlorideH-Lys(Z)-OBzl·HClH-Lys(Z)-OBzl.HCl

0420175GM
100292-490EA 133.38 USD
100292-490
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥99% (by HPLC), crystalline powder
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₂₁H₂₆N₂O₄·HCl
MW: 406.9 g/mol
Melting Pt: 136…141 °C
MDL Number: MFCD00038981
CAS Number: 6366-70-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/12/21286612.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR