Flavokawain A ≥98%, powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
2'-Hydroxy-4,4',6'-trimethoxychalcone
, Flavokawain A
, 1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
021043100MG
100266-046EA
117.69
USD
100266-046
Flavokawain A ≥98%, powder
Flavokawain A
- Yellow powder
- Store in a cool, dry place
Formula:
C₁₈H₁₈O₅ MW: 314.34 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD00017174 CAS Number: 3420-72-2 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...