Histidylphenylalanine

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-His-Phe-OH

0417791GM
100292-388EA 1085.38 USD
100292-388
Histidylphenylalanine
Histidylphenylalanine
Formula: C₁₅H₁₈N₄O₃
MW: 302.334 g/mol
MDL Number: MFCD00037858
CAS Number: 16874-81-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/01/21287201.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR