2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1H,1H-Perfluoro-1-butanol2,2,3,3,4,4,4-Heptafluorobutan-1-olHeptafluoro-1-butanol

09141025GM
100274-466EA 197.46 USD
100274-466
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 95…96.5 °C
MDL Number: MFCD00004674
CAS Number: 375-01-9
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR