n-Heptacosane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Heptacosane

05270025MG
100270-572EA 258.92 USD
100270-572
n-Heptacosane ≥99%
n-Heptacosane
Formula: H₃C(CH₂)₂₅CH₃
MW: 380.74 g/mol
MDL Number: MFCD00009862
CAS Number: 593-49-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/85/21285985.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR