Fmoc-Glu(OcHx)-OH ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
N-Fmoc-L-glutamic acid 5-cyclohexyl ester
, Fmoc-Glu(OcHex)-OH
041275GM
100278-982EA
58.85
USD
100278-982
Fmoc-Glu(OcHx)-OH ≥99%
Fmoc-Glu(OcHx)-OH
Formula:
C₂₆H₂₉NO₆ MW: 451.519 g/mol |
MDL Number:
MFCD00270538 CAS Number: 150047-85-1 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...