3,4-Dinitrofluorobenzene ≥97%, crystalline solid
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
4-Fluoro-1,2-dinitrobenzene
213171GM
100293-632EA
137.31
USD
100293-632
3,4-Dinitrofluorobenzene ≥97%, crystalline solid
3,4-Dinitrofluorobenzene
- Crystalline solid
- Store in a cool, dry place
Formula:
C₆H₃FN₂O₄ MW: 186.1 g/mol Melting Pt: 55…56 °C |
MDL Number:
MFCD00236671 CAS Number: 364-53-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...