Cytochalasin A, powder
Supplier: INDOFINE CHEMICAL COMPANY MS
0215611MG
100268-676EA
289
USD
100268-676
Cytochalasin A, powder
Cytochalasin A
- Powder
- Store in a cool, dry place
Formula:
C₂₉H₃₅NO₅ Melting Pt: 191…193 °C |
MDL Number:
MFCD00005935 CAS Number: 14110-64-6 UN: 1544 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...