L(+)-Cysteine hydrochloride monohydrate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate(R)-(+)-Cysteine hydrochloride monohydrateL-Cysteine HCI MonohydrateCysteine Hydrochloride monohydrateH-Cys-OH.HCl.H2OCysteine Hydrochloride, Monohydrate

041081250GM
100278-704EA 129.46 USD
100278-704
L(+)-Cysteine hydrochloride monohydrate ≥99%
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
MDL Number: MFCD00065606
CAS Number: 7048-04-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/13/21286513.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR