Cycloartenol acetate ≥90% (by HPLC)
Supplier: INDOFINE CHEMICAL COMPANY MS
0200115MG
100269-674EA
640.77
USD
100269-674
Cycloartenol acetate ≥90% (by HPLC)
Cycloartenol acetate
Formula:
C₃₂H₅₂O₂ |
MDL Number:
MFCD00200960 CAS Number: 1259-10-5 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...