Citral ≥95%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
3,7-Dimethylocta-2,6-dienal
025016S1GM
100263-686EA
243.23
USD
100263-686
Citral ≥95%
Citral
Formula:
C₁₀H₁₆O MW: 152.24 g/mol |
MDL Number:
MFCD00006997 CAS Number: 5392-40-5 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...