Boc-D-Ser-OBzl ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-D-serine benzyl ester

048721GM
100277-440EA 222.31 USD
100277-440
Boc-D-Ser-OBzl ≥99% (by HPLC), powder
Boc-D-Ser-OBzl

  • White to off-white powder
  • Store at 0-8 °C

Formula: C₁₅H₂₁NO₅
Melting Pt: 66…70 °C
MDL Number: MFCD00080265
CAS Number: 141527-78-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/48/21286448.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR