Boc-D-3-Pyridylalanine ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-β-(3-pyridyl)-D-Ala-OHBoc-D-3-Pal-OHBoc-3-(3-pyridyl)-D-Ala-OH

048661GM
100281-928EA 166.08 USD
100281-928
Boc-D-3-Pyridylalanine ≥98% (by HPLC), powder
Boc-D-3-Pyridylalanine

  • White powder
  • Store at 0-8 °C

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
Melting Pt: 134…148 °C
MDL Number: MFCD00076901
CAS Number: 98266-33-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/47/21286447.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR