Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-D-Lys-OHNɑ-Boc-D-lysine

047631GM
100281-770EA 85 USD
100281-770
Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98% (by HPLC), powder
Nalpha-(tert-Butoxycarbonyl)-D-lysine

  • White powder
  • Store at 0-8 °C

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
Melting Pt: 192…212 °C
MDL Number: MFCD00076956
CAS Number: 106719-44-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/41/21287241.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR