Extra-Safe™ Hip Length Labcoats, ValuMax®
Supplier: Valumax International, Inc.
3630CB2XL 3630CYS 3630CBXL 3630CBL 3630CBM 3630CYXS
10497-974CS 625.2 USD
10497-974 10497-980 10497-982 10497-984 10497-986 10498-214
Extra-Safe™ Hip Length Labcoats, ValuMax®
Disposable Garments
Durable unisex lab jacket made for protection and comfort with SMS material that is fluid resistant, highly breathable, and fire retardant.
  • Soft, protective 3-layer SMS fabrics
  • Fluid-resistant and breathable
  • Autoclavable & washable up to 5 times
  • Latex-free & fire retardant
  • Anti-static – no clinging
These lab coats are available in 30 colors with knit cuffs and collar.
Certifications: Meets OSHA requirements.