Clostridium histolyticum Collagenase
Supplier: Thermo Fisher Scientific
J13820-03 J13820-06
101174-296EA 400.02 USD
101174-296 101170-394
Clostridium histolyticum Collagenase
Enzymes
Collagenase contains a mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes.
  • Enzyme activity: ≥125 units/mg powder (dry basis)
  • Enzyme activity: ≥0.15 Wunsch units/mg (powder dry basis)
One unit liberates 1 μmol of L-Leucine equivalents from collagen in 5 hr at 37 °C, pH 7.5. One unit is equal to 0.0012 Wunsch units.