Microflex® L91 Latex Extended Cuff Examination Gloves, Ansell
Supplier: Ansell Healthcare
L912 L913
10500-598CS 447.95 USD
10500-598 10500-600 10500-634 10500-636
Microflex® L91 Latex Extended Cuff Examination Gloves, Ansell
Gloves
These extended cuff latex examination gloves are thicker and stronger than standard exam gloves to give the wearer extra protection in hazardous environments.
  • Beaded cuff and a textured finish for excellent gripping power
  • Longer length for added protection
Certifications: 510K approved for medical use
Packaging: Gloves are packaged 50 gloves in a box; 10 boxes in a carton.