Vacuum Traps, O-Ring Joints, Chemglass
Supplier: Chemglass
CG-4513-10 CG-4513-11 CG-4513-12 CG-4513-20 CG-4513-21 CG-4513-22 CG-4513-30 CG-4513-31 CG-4513-32
89029-410EA 450.64 USD
89029-410 89029-412 89029-414 89029-416 89029-418 89029-420 89029-422 89029-424 89029-426
Vacuum Traps, O-Ring Joints, Chemglass
Vacuum Traps
Vacuum traps with an o-ring joint to connect the trap body to the stopper
O-ring joint provides for a vacuum tight, grease free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Serrated hose connections are 13.5 mm at the smallest serration and 15.5 mm at the largest serration.
Delivery information: Complete trap is supplied with a stopper, lower body, viton o-ring and stainless steel pinch clamp.