Aaron RAM™ Low Temperature Cautery Pens, Stoelting
Supplier: Stoelting
59117 59119 59015
95060-234EA 187.82 USD
95060-234 95060-242 95060-238
Aaron RAM™ Low Temperature Cautery Pens, Stoelting
Pens Surgical Markers
With a variety of low temperatures and fine tips, the surgeon is assured of precise pinpoint hemostasis.
  • Disposable
  • Four-year shelf life
Packaging: Individually packaged, sterile, 10 per box.