Lab Frame Foot, Chemglass
Supplier: Chemglass
CG-9274-01 CG-9274-S-01
80063-668EA 49.54 USD
80063-668 89094-302
Lab Frame Foot, Chemglass
Support Rods and Frames
Lab frame feet in two different versions.
  • Permanent lattice mounting to bench tops, floors or walls
  • Accommodates 1/2” (13 mm) diameter rod
  • Available in stainless steel electro-polished finish
Supplied with three screws.