T7 RNA Polymerase, Promega
Supplier: Promega Corporation
P2075
P2077
PAP2075EA
178.03
USD
PAP2075
PAP2077
T7 RNA Polymerase, Promega
Nucleic Acid Reagents
RNA Synthesis and Transcription
DNA-dependent RNA polymerase for in vitro transcription.
- DNA-Dependent RNA Polymerase Specific for T7 Promoter Sequences
- >90% pure as determined by SDS polyacrylamide gel electrophoresis
- Free of detectable levels of contaminating RNase and DNase activity (
- Incorporates 32P, 33P, 3H and 35S nucleoside triphosphates
T7 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only T7 DNA or DNA cloned downstream from a T7 promoter can serve as a template for T7 RNA Polymerase-directed RNA synthesis.