Glass Tubing Cutter, Humboldt
Supplier: Humboldt
H-13550
72410-038EA 489.7 USD
72410-038
Glass Tubing Cutter, Humboldt
Cutters
Aluminum alloy casting with gray hammerloid finish.
  • Attaches to laboratory table tops up to 45 mm thick
  • Cuts glass tubing up to 11/2" (38 mm) diameter
Hardened steel cutting wheel.