n-Hexane 95, CHROMASOLV™ Pest_Residual, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

34484-4X4L 34484-1L 34484-4X2.5L
BJ34484-4LCS 1205.15 USD
BJ34484-4L BJ34484-1L BJ34484-2.5L BJ34484-4X2.5L
n-Hexane 95, CHROMASOLV™ Pest_Residual, Riedel-de Haën™
n-Hexane

Hexane (C6H14) is a flammable colorless liquid; it has a faint odor and is highly volatile. It is a chief constituent of petroleum, petroleum ether and gasoline. It is used as a solvent for adhesives, vegetable oils and to extract oil and grease contaminants from water and soil for analysis. Hexanes are commonly used in chromatography as a non-polar solvent. Suitable for GC and Pesticide Residue Analysis.


  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.660…0.668 g/cm³ (20 °C) (lit.)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 95 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Autoignition Temperature 453 °F
Explosion Limit 7.7 %
Vapor Pressure ~132 mmHg ( 20 °C)
Determination of P/N ≤5 ng/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR