Qualaknit® ESD Nylon Conductive Knit ESD Gloves, QRP
Supplier: QRP
Qualaknit®
KASM KASS KASL KASXL
32888-997CS 647.19 USD
32888-997 32884-911 32888-993 32888-991
Qualaknit® ESD Nylon Conductive Knit ESD Gloves, QRP
Gloves
The QRP KAS Qualaknit ESD Assembly/Inspection Gloves are uncoated gloves for maximum breathability and lowest resistance.
  • Nylon + conductive carbon/nylon yarn
  • Micro-knit fingertips for maximum dexterity
  • Seamless knit for greater comfort
  • Surface resistivity 10⁸ ohms
KAS Qualaknit ESD Assembly-Inspection Glove is knit from continuous stretch nylon and carbon filiment fibers.