Qualatrile® Indy BQY09 Qualatrile Indy Nitrile 9" Powder-Free Gloves, QRP
Supplier: PIP
BQY09M BQY09XL
89041-010CS 438.26 USD
89041-010 89041-012 89041-016
Qualatrile® Indy BQY09 Qualatrile Indy Nitrile 9" Powder-Free Gloves, QRP
Gloves
The QRP BQY09 gloves are industrial grade, RoHS compliant, ambidextrous, powder-free gloves that are 100% nitrile.
  • 4 mil - 3× puncture resistance of vinyl or natural rubber latex gloves
  • Tough industrial grade
  • Designed for automotive applications
  • Economical protection from chemicals and petroleum
  • USDA compliant for food handling and processing
  • Naturally anti-static, surface resistance 10⁹ ohms
  • Distinctive blue color