innuPREP Gel Extraction Kit, Analytik Jena
10755-620EA 128.55 USD
10755-620 10755-784
innuPREP Gel Extraction Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The innuPREP Gel Extraction Kit is a tool for extremely fast, simple isolation and concentration of DNA fragments from TAE or TBE agarose gels.
  • Fast, efficient isolation of DNA from TAE or TBE agarose gels (up to 300mg)
  • High recovery rates and excellent quality for subsequent applications
  • Capable of processing a large range of fragment lengths (from 100bp to 30kb)
  • Elution volumes can be reduced to 10µl
The innuPREP Gel Extraction Kit is a tool for extremely fast, simple isolation and concentration of DNA fragments from TAE or TBE agarose gels. The first step of the process on which the kit is based is to solubilize agarose gel pieces. This is then followed by selective binding of the DNA onto a filter membrane. The bound DNA is then washed and desorbed from the filter membrane by adding a low-salt buffer. The elution volumes used in this process may vary between 10µl and 50µl. All of the buffers have been adjusted to work well together, resulting in a highly efficient recovery process. The isolated DNA fragments are suitable for immediate use in further applications.