Adhesive Vinyl GHS OSHA Labels 'Ozone', HCL Labels
Supplier: HCL Label
GH1X570VN0023 GH1X570VN0035 GH1X570VN0047 GH2X570VN0023 GH2X570VN0035 GH2X570VN0047 GH3X570VN0023 GH3X570VN0035 GH3X570VN0047 GH4X570VN0035 GH4X570VN0047 GH4X570VN0023
103026-570PK 74.03 USD
103026-570 103026-572 103026-574 103026-576 103026-578 103026-580 103034-588 103034-590 103034-592 103034-594 103034-596 103034-598 103041-632 103041-634 103041-636 103041-638 103041-640 103041-642 103050-594 103050-596 103050-598 103050-600 103050-686 103050-688
Adhesive Vinyl GHS OSHA Labels 'Ozone', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.