Adhesive Vinyl GHS OSHA Labels 'Nickel Oxide', HCL Labels
Supplier: HCL Label
GH11734VN0023 GH11734VN0035 GH11734VN0047 GH21734VN0023 GH21734VN0035 GH21734VN0047 GH31734VN0023 GH31734VN0035 GH31734VN0047 GH41734VN0023 GH41734VN0035 GH41734VN0047
103025-174PK 74.03 USD
103025-174 103025-176 103025-178 103025-180 103025-182 103025-184 103033-120 103033-122 103033-124 103033-126 103033-128 103033-130 103040-676 103040-678 103041-598 103041-600 103041-602 103041-604 103048-260 103048-262 103048-264 103048-266 103048-268 103048-270
Adhesive Vinyl GHS OSHA Labels 'Nickel Oxide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.