Adhesive Vinyl GHS OSHA Labels 'Phenolphthalein', HCL Labels
Supplier: HCL Label
GH11731VN0023 GH11731VN0035 GH11731VN0047 GH21731VN0023 GH21731VN0035 GH21731VN0047 GH31731VN0023 GH31731VN0035 GH31731VN0047 GH41731VN0035 GH41731VN0047 GH41731VN0023
103025-162PK 74.03 USD
103025-162 103025-164 103025-166 103025-168 103025-170 103025-172 103033-108 103033-110 103033-112 103033-114 103033-116 103033-118 103040-608 103040-610 103040-668 103040-670 103040-672 103040-674 103048-252 103048-254 103048-256 103048-258 103049-038 103049-040
Adhesive Vinyl GHS OSHA Labels 'Phenolphthalein', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.