Adhesive Vinyl GHS OSHA Labels 'Sodium Sulfate', HCL Labels
Supplier: HCL Label
GH11601VN0023 GH11601VN0035 GH11601VN0047 GH21601VN0023 GH21601VN0035 GH21601VN0047 GH31601VN0023 GH31601VN0035 GH31601VN0047 GH41601VN0023 GH41601VN0035 GH41601VN0047
103024-818PK 74.03 USD
103024-818 103024-820 103024-822 103024-824 103024-826 103024-828 103032-820 103032-822 103032-824 103032-826 103032-828 103032-830 103041-344 103041-346 103041-348 103041-350 103041-352 103041-354 103048-056 103048-058 103048-060 103048-062 103048-064 103048-066
Adhesive Vinyl GHS OSHA Labels 'Sodium Sulfate', HCL Labels
Labels
These ready-made laminated adhesive vinyl labels comply with the updated OSHA HazCom secondary container labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.