Adhesive Vinyl GHS OSHA Labels 'Aluminum Oxide', HCL Labels
Supplier: HCL Label
GH11415VN0023 GH11415VN0035 GH11415VN0047 GH21415VN0023 GH21415VN0035 GH21415VN0047 GH31415VN0023 GH31415VN0035 GH31415VN0047 GH41415VN0023 GH41415VN0035 GH41415VN0047
103024-430PK 74.03 USD
103024-430 103024-432 103024-570 103024-572 103024-574 103024-576 103032-424 103032-426 103032-428 103032-430 103032-512 103032-514 103040-964 103040-966 103040-968 103040-970 103040-972 103040-974 103047-780 103047-782 103047-784 103047-786 103047-788 103047-790
Adhesive Vinyl GHS OSHA Labels 'Aluminum Oxide', HCL Labels
Labels
These ready-made laminated adhesive vinyl labels comply with the updated OSHA HazCom secondary container labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS). All sizes are protected with a chemical resistant lamination and a shelf/outdoor life of two years.
Certifications: RoHS compliant.