Adhesive Vinyl GHS OSHA Labels 'Hexachlorobutadiene', HCL Labels
Supplier: HCL Label
GH10513VN0023 GH10513VN0035 GH10513VN0047 GH20513VN0023 GH20513VN0035 GH20513VN0047 GH30513VN0023 GH30513VN0035 GH30513VN0047 GH40513VN0023 GH40513VN0035 GH40513VN0047
103022-438PK 74.03 USD
103022-438 103022-440 103022-442 103022-444 103022-446 103022-448 103030-048 103030-050 103030-052 103030-054 103030-056 103030-058 103037-808 103037-810 103037-812 103037-814 103037-816 103037-818 103045-636 103045-638 103045-640 103045-642 103045-644 103045-646
Adhesive Vinyl GHS OSHA Labels 'Hexachlorobutadiene', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.