Adhesive Vinyl GHS OSHA Labels 'Gallium Trichloride', HCL Labels
Supplier: HCL Label
GH10507VN0023 GH10507VN0035 GH10507VN0047 GH20507VN0047 GH20507VN0023 GH20507VN0035 GH30507VN0047 GH30507VN0023 GH30507VN0035 GH40507VN0023 GH40507VN0035 GH40507VN0047
103022-366PK 74.03 USD
103022-366 103022-368 103022-370 103022-372 103022-374 103022-376 103029-488 103029-490 103030-016 103030-018 103030-020 103030-022 103037-704 103037-706 103038-176 103038-178 103038-180 103038-182 103045-596 103045-598 103045-600 103045-602 103045-604 103048-612
Adhesive Vinyl GHS OSHA Labels 'Gallium Trichloride', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.