Adhesive Vinyl GHS OSHA Labels 'Diesel Fuel', HCL Labels
Supplier: HCL Label
GH10324VN0023 GH10324VN0035 GH10324VN0047 GH20324VN0023 GH20324VN0035 GH20324VN0047 GH30324VN0035 GH30324VN0047 GH30324VN0023 GH40324VN0023 GH40324VN0035 GH40324VN0047
103020-674PK 74.03 USD
103020-674 103020-676 103020-678 103020-680 103020-682 103020-684 103028-098 103028-100 103028-102 103028-104 103028-106 103028-108 103036-464 103036-466 103036-468 103036-470 103036-828 103036-830 103044-464 103044-466 103044-468 103044-470 103044-472 103044-474
Adhesive Vinyl GHS OSHA Labels 'Diesel Fuel', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.