Adhesive Vinyl GHS OSHA Labels 'Butyl Alcohol', HCL Labels
Supplier: HCL Label
GH10220VN0023 GH10220VN0035 GH10220VN0047 GH20220VN0023 GH20220VN0035 GH20220VN0047 GH30220VN0023 GH30220VN0035 GH30220VN0047 GH40220VN0023 GH40220VN0035 GH40220VN0047
103020-206PK 74.03 USD
103020-206 103020-208 103020-210 103020-212 103020-214 103020-216 103028-054 103028-056 103028-058 103028-060 103028-062 103028-064 103036-376 103036-378 103036-380 103036-382 103036-384 103036-386 103043-972 103043-974 103043-976 103043-978 103043-980 103043-982
Adhesive Vinyl GHS OSHA Labels 'Butyl Alcohol', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.