Adhesive Vinyl GHS OSHA Labels 'Ammonium Fluoride', HCL Labels
Supplier: HCL Label
GH10037VN0023 GH10037VN0035 GH10037VN0047 GH20037VN0023 GH20037VN0035 GH20037VN0047 GH20038VN0023 GH20038VN0035 GH30037VN0023 GH30037VN0035 GH30037VN0047 GH30038VN0023 GH30038VN0035 GH40037VN0023 GH40037VN0035 GH40037VN0047 GH40038VN0023 GH40038VN0035
103019-480PK 74.03 USD
103019-480 103019-482 103019-484 103019-486 103019-488 103019-490 103027-368 103027-370 103027-372 103027-374 103027-376 103027-378 103027-380 103027-382 103027-384 103035-786 103035-788 103035-790 103035-792 103035-794 103035-796 103035-798 103035-800 103035-802 103043-238 103043-240 103043-242 103043-284 103043-286 103043-288 103043-290 103043-292 103043-294
Adhesive Vinyl GHS OSHA Labels 'Ammonium Fluoride', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant.