Eco Best Technology® Nitrile Industrial Gloves, Powder-Free, Showa
Supplier: Avantor
728-09
75804-772CS 732.79 USD
75804-772 75804-774 75804-776 75804-778 75804-780 75804-782
Eco Best Technology® Nitrile Industrial Gloves, Powder-Free, Showa
Gloves
Using revolutionary Eco Best Technology®, this mid-forearm gauntlet is designed for multiple uses and a range of applications, from food processing to manufacturing, masonry to refinery operations.
  • 100% nitrile with Eco Best Technology®
  • Unsupported, unlined
  • 15 mil thick
  • Bisque grip
These nitrile gloves provide chemical protection against solvents and acids, and their bisque finish grip makes wet work easier and safer. Effective gripping allows superior tactile precision.