Supplied Air Respirator Parts & Accessories, 3M™, 3M Company
Supplier: 3M
7000051867 1010015234 7000051868 7000051866
67101-364CS 187.45 USD
67101-364 67101-272 67101-362 67101-366
Supplied Air Respirator Parts & Accessories, 3M™, 3M Company
Respirators Respiratory Systems
An assortment of replacement parts and accessories designed for use on a variety of 3M™ Supplied Air Respirators.
Wing Nut W-2916 is a component of the 3M™ Compressed Air Filter and Regulator Panel W-2806.
High pressure Socket W-3281-2 is designed for use on the 3M™ Compressed Air Filter and Regulator Panel W-2806. It has a brass finish. High pressure Hose Socket W-3280-2 may be used on the 3M™ Supplied Air Hose W-9435 and W-2929. It also has a brass finish.
High pressure hose plug W-3279-2 may be used on the 3M™ Supplied Air Hose W-9435 or W-2929. It has a brass finish.
Neck protectors areMore Product Information