PvuI
Supplier: Promega Corporation
R6321
R6325
PAR6321EA
108.72
USD
PAR6321
PAR6325
PvuI
Nucleic Acid Reagents
Restriction Enzymes
CG AT/CG GC\TA GC
- Active and capable of digestion directly in GoTaq(R) Green Master Mix