Human Peripheral Blood Naïve CD4 Helper T Cells, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB0445RAC-1
103051-092EA 1234.62 USD
103051-092
Human Peripheral Blood Naïve CD4 Helper T Cells, Charles River Laboratories Cell Solutions
Cells
Naïve CD4 T cells are isolated from a LeukoPak collected from IRB-consented volunteer donors.
  • Isolated from mononuclear cells by positive selection
  • Negative selection is also available
  • Immunomagnetic cell separation procedures
Naïve CD4 T cells have differentiated from bone marrow, and have successfully undergone the central tolerance processes of positive and negative selection in the thymus
Naïve CD4+ T Cells are isolated from mononuclear cells by positive selection using immunomagnetic cell separation procedures. Negative selection is also available at your request.
They are considered mature but inactivated, responding to novel pathogens in the immune system and initiate the immune response. They are used to illicit brand new immune responses in in vitro and in vivo studies.