Hematoxylin, powder, Surgipath®, certified by the Biological Stain Commission
Supplier: Leica Biosystems
Synonyms:
(6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol
, Hydroxybrasilin
, NSC 2785
, DcH-48
, Hydroxybrazilin
, (+)-Hematoxylin
, Natural Black I
Powdered Stain Hematoxylin Dye can be used to prepare all Hematoxylin formulations for routine and special staining procedures that require nuclear staining.
- Certified by the Biological Stain Commission
- Colour Index Number 75290
The total dye content is stated on each label.
Certifications: Dye is certified by the Biological Stain Commission.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...