Surgipath® Prefilled Formalin Specimen Jars, Leica
Supplier: Leica Biosystems
3800759 3800780 3800790 3800810 3800840 380077027 380075432AS
75810-266CS 822.91 USD
75810-266 75810-268 75810-270 75810-272 75810-276 75810-294 75810-296 75810-298
Surgipath® Prefilled Formalin Specimen Jars, Leica
Jars Specimen Jars
Pre-filled Formalin Specimen Jars are impact-resistant, clear polypropylene jars prefilled with 10% Neutral, Phosphate Buffered Formalin.
  • Wide mouth
  • Ribbed caps
  • Leakproof
  • Great for small specimens
  • Easy to store and ship
Wide-mouth jars provide easy access to specimens
AffirmSeal jars are made of durable plastic to prevent cracking when inadvertently dropped, and comply with 95 kPa pressure testing for safe air shipment and use in pneumatic transport systems.
The amount of formalin in the specimen jars is approximately half the volume of the jar. Caps are ribbed, leakproof and easy to open or close even with wet hands or gloves. Lid provides an audible and tactile acknowledgement when jar is closed properly.