Surgipath® Stainless Steel Base Molds, Leica
Supplier: Leica
Surgipath®
3803081 3803080 3803082 3803084 3803083
75809-390PK 234.98 USD
75809-390 75809-392 75809-384 75809-386 75809-388
Surgipath® Stainless Steel Base Molds, Leica
Moulds Moulds for Histology
These re-usable base molds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
  • Available in 5 sizes
  • Reusable
  • Internal rounded corners
  • Easy block removal
These base molds can be used on all standard size cassettes.
Packaging: Molds are sold in packs of 12.