Glycerine solution 80%, sterile

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

113055044 113055034
IC113055044EA 196.7 USD
IC113055044 76019-966
Glycerine solution 80%, sterile
Glycerine

Sterile 80% Glycerol solution offered is less viscous and more convenient to pipette than 100% glycerol.


Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component. It has been used in purification of bacteriophage λ particles by step gradients, calcium-phosphate-mediated transfection of eukaryotic cells with plasmid DNA, DEAE-dextranmediated transfection of DNA into cells, DNA sequencing reactions, and mediation of mispriming in PCR. The use of glycerol in improved focusing of alkaline pH protein samples in immobilized pH gradient (IPG) strips has been published. Studies of the effect of glycerol on separation of DNA fragments by capillary electrophoresis have been reported.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 182 °C (20 mmHg)) (lit.)
Melting Pt: 18.6
Density: 1.25 g/mL (lit.)
Flash Pt: 177
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals ≤2 ppm
Water Content ≤0.5%
Storage Store at Room Temperature (15-30 °C)
Assay ≥99.5% (GC), corrected for water
Concentration 0.8
Auto Ignition 698 °F (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR