Adenine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7H-Purin-6-amine6-AminopurinePurin-6-amine

02700125GM
100266-524EA 204 USD
100266-524
Adenine ≥99%
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: >360 °C (dec.)
MDL Number: MFCD00041790
CAS Number: 73-24-5
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/57/19/21285719.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR