Xylene (mixture of isomers) ACS

Supplier: AQUA SOLUTIONS MS
Warning

Synonyms: Dimethylbenzene

X7502-4L X7502-4L(PVC)
101225-690EA 194.75 USD
101225-690 101225-802
Xylene (mixture of isomers) ACS
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥98.5%
Color (APHA) ≤10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR